Product Details

SNP ID
rs199710268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29063420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGCCACTTTGCCACCTGTTAC[C/T]ACGACATTTGCACCAGTATCAGCAA
Phenotype
MIM: 604735
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCT8 PubMed Links

Gene Details

Gene
CCT8
Gene Name
chaperonin containing TCP1 subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282907.1 Intron NP_001269836.1
NM_001282908.1 Intron NP_001269837.1
NM_001282909.1 Intron NP_001269838.1
NM_006585.3 Intron NP_006576.2
Gene
USP16
Gene Name
ubiquitin specific peptidase 16
There are no transcripts associated with this gene.

View Full Product Details