Product Details

SNP ID
rs200025631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41740897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCATGGCCGCCCTGGAACTTGA[C/G]GCTCTGCAGGTTGATGTGGGCCCCA
Phenotype
MIM: 605706
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6814 PubMed Links

Gene Details

Gene
MIR6814
Gene Name
microRNA 6814
There are no transcripts associated with this gene.

Gene
RIPK4
Gene Name
receptor interacting serine/threonine kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020639.2 2344 Missense Mutation CTC,GTC L766V NP_065690.2

View Full Product Details