Product Details

SNP ID
rs201528567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30490246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGTGTTGTCGAAGCGCAGGACA[A/T]CTGCAGTGATGGAGAGGTGATCAGG
Phenotype
MIM: 612825
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SDC4P PubMed Links

Gene Details

Gene
SDC4P
Gene Name
syndecan 4 pseudogene
There are no transcripts associated with this gene.

Gene
SEC14L4
Gene Name
SEC14 like lipid binding 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161368.1 984 Intron NP_001154840.1
NM_174977.3 984 Missense Mutation TAT,TTT Y361F NP_777637.1
XM_006724235.2 984 Missense Mutation TAT,TTT Y307F XP_006724298.1
XM_011530164.2 984 Missense Mutation TAT,TTT Y346F XP_011528466.1
XM_017028787.1 984 Missense Mutation TAT,TTT Y420F XP_016884276.1
XM_017028788.1 984 Missense Mutation TAT,TTT Y392F XP_016884277.1
XM_017028789.1 984 Missense Mutation TAT,TTT Y307F XP_016884278.1
XM_017028790.1 984 Intron XP_016884279.1
XM_017028791.1 984 Missense Mutation TAT,TTT Y307F XP_016884280.1

View Full Product Details