Product Details

SNP ID
rs199556141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:45672118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTCGGGCGTCATGGTGCCGGCG[C/T]CCATCCAAGACCTGGAGGCCCTGCG
Phenotype
MIM: 611150
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATXN10 PubMed Links

Gene Details

Gene
ATXN10
Gene Name
ataxin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167621.1 321 Missense Mutation CCC,TCC P19S NP_001161093.1
NM_013236.3 321 Missense Mutation CCC,TCC P19S NP_037368.1

View Full Product Details