Product Details

SNP ID
rs200674632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36227430 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGGTCTTGGCCTGGCTGCAGCA[G/T]CTCATGGATCTTGGTGAGAAAGTTG
Phenotype
MIM: 607252
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
APOL2 PubMed Links

Gene Details

Gene
APOL2
Gene Name
apolipoprotein L2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030882.3 1465 Intron NP_112092.2
NM_145637.2 1465 Missense Mutation ATG,CTG M330L NP_663612.2
XM_011530074.1 1465 Intron XP_011528376.1
XM_011530075.1 1465 Intron XP_011528377.1
XM_011530076.2 1465 Intron XP_011528378.1
XM_011530077.2 1465 Intron XP_011528379.1
XM_011530078.1 1465 Intron XP_011528380.1
XM_017028724.1 1465 Intron XP_016884213.1

View Full Product Details