Product Details

SNP ID
rs200736981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40832642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTTACAGGCAAGGGCAAGATCA[C/T]GGGCTGCTTCTTCCCAGTGGCCTAG
Phenotype
MIM: 606796
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ST13 PubMed Links

Gene Details

Gene
ST13
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278589.1 1076 Missense Mutation CAT,CGT H193R NP_001265518.1
NM_003932.4 1076 Missense Mutation CAT,CGT H203R NP_003923.2

View Full Product Details