Product Details

SNP ID
rs201003382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19432575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCATGACGGCCTCCGTGCTGCG[A/C]AGTATCTCGCTAGCCCTGCGCCCGA
Phenotype
MIM: 600237 MIM: 605089
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C22orf39 PubMed Links

Gene Details

Gene
C22orf39
Gene Name
chromosome 22 open reading frame 39
There are no transcripts associated with this gene.

Gene
HIRA
Gene Name
histone cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003325.3 674 Intron NP_003316.3
Gene
MRPL40
Gene Name
mitochondrial ribosomal protein L40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318151.1 674 UTR 5 NP_001305080.1
NM_001318152.1 674 Intron NP_001305081.1
NM_003776.3 674 Silent Mutation CGA,CGC R7R NP_003767.2

View Full Product Details