Product Details

SNP ID
rs201207008
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22244992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATCAGCCGCCGGCCATGTCCTC[A/G]GCCCTTGGAACCACAATCCGCCTCA
Phenotype
MIM: 605141
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
VPREB1 PubMed Links

Gene Details

Gene
VPREB1
Gene Name
pre-B lymphocyte 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303509.1 113 Silent Mutation TCA,TCG S30S NP_001290438.1
NM_007128.3 113 Silent Mutation TCA,TCG S31S NP_009059.1

View Full Product Details