Product Details

SNP ID
rs201630786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23753105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGACACACCGTAGTCCCTGATG[G/T]TGACGTGCTGGGGGCTGAGCGTGCA
Phenotype
MIM: 605017 MIM: 194544
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C22orf15 PubMed Links

Gene Details

Gene
C22orf15
Gene Name
chromosome 22 open reading frame 15
There are no transcripts associated with this gene.

Gene
VPREB3
Gene Name
pre-B lymphocyte 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013378.2 223 Missense Mutation AAC,ACC N48T NP_037510.1
Gene
ZNF70
Gene Name
zinc finger protein 70
There are no transcripts associated with this gene.

View Full Product Details