Product Details

SNP ID
rs200680599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20116057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCACTTGGCAAGTTTCTGCAGCA[C/T]CTGCTCGCACTCCAGCTGCTTCCGC
Phenotype
MIM: 609030 MIM: 601180 MIM: 611151
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGCR8 PubMed Links

Gene Details

Gene
DGCR8
Gene Name
DGCR8 microprocessor complex subunit
There are no transcripts associated with this gene.

Gene
MIR6816
Gene Name
microRNA 6816
There are no transcripts associated with this gene.

Gene
RANBP1
Gene Name
RAN binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278639.1 776 UTR 5 NP_001265568.1
NM_001278640.1 776 Intron NP_001265569.1
NM_001278641.1 776 Intron NP_001265570.1
NM_002882.3 776 Intron NP_002873.1
XM_011530289.2 776 Intron XP_011528591.1
XM_011530290.2 776 Intron XP_011528592.1
XM_011530291.2 776 Intron XP_011528593.1
XM_017028890.1 776 Intron XP_016884379.1
XM_017028891.1 776 Intron XP_016884380.1
XM_017028892.1 776 Intron XP_016884381.1
Gene
TRMT2A
Gene Name
tRNA methyltransferase 2 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257994.1 776 Missense Mutation ATG,GTG M194V NP_001244923.1
NM_022727.5 776 Missense Mutation ATG,GTG M194V NP_073564.3
NM_182984.4 776 Missense Mutation ATG,GTG M194V NP_892029.2
XM_011530139.2 776 Missense Mutation ATG,GTG M194V XP_011528441.1
XM_011530141.1 776 UTR 5 XP_011528443.1
XM_011530142.2 776 Intron XP_011528444.1
XM_017028770.1 776 Missense Mutation ATG,GTG M194V XP_016884259.1

View Full Product Details