Product Details

SNP ID
rs200811246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40861441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTTGTCTGAAGTTGTAACAGAT[A/G]TGTAGTTGTCCATCCCACTATTATC
Phenotype
MIM: 611336 MIM: 606796 MIM: 613553
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB7 PubMed Links

Gene Details

Gene
DNAJB7
Gene Name
DnaJ heat shock protein family (Hsp40) member B7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145174.1 686 Missense Mutation ACA,ATA T185I NP_660157.1
Gene
ST13
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein)
There are no transcripts associated with this gene.

Gene
XPNPEP3
Gene Name
X-prolyl aminopeptidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204827.1 686 Intron NP_001191756.1
NM_022098.3 686 Intron NP_071381.1

View Full Product Details