Product Details

SNP ID
rs201706183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19450741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCGACGAATCTGCCTCCAGCTT[C/T]ATCCTAGGTTTGAAGAGAAGATACT
Phenotype
MIM: 601754
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C22orf39 PubMed Links

Gene Details

Gene
C22orf39
Gene Name
chromosome 22 open reading frame 39
There are no transcripts associated with this gene.

Gene
UFD1L
Gene Name
ubiquitin fusion degradation 1 like (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035247.2 1109 Intron NP_001030324.2
NM_005659.6 1109 Intron NP_005650.2
XM_011530361.2 1109 Missense Mutation AAA,GAA K280E XP_011528663.1

View Full Product Details