Product Details
- SNP ID
-
rs199649182
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:50482734 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGGCCGCCAACACTCGGGCCCCC[A/G]AAGACACTCGGGCCCCCGCAGGACC
- Phenotype
-
MIM: 608682
MIM: 606774
MIM: 603560
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADM2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72438078] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADM2
- Gene Name
- adrenomedullin 2
- Gene
- MIOX
- Gene Name
- myo-inositol oxygenase
There are no transcripts associated with this gene.
- Gene
- SBF1
- Gene Name
- SET binding factor 1
There are no transcripts associated with this gene.
View Full Product Details