Product Details

SNP ID
rs200684011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41603339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGAACTGTGCCACTTCGTTGCT[A/G]AAGGTGTTACAATTGTGTTCAAAGA
Phenotype
MIM: 614637
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DESI1 PubMed Links

Gene Details

Gene
DESI1
Gene Name
desumoylating isopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015704.2 571 Silent Mutation TTC,TTT F111F NP_056519.1
XM_005261571.4 571 Silent Mutation TTC,TTT F111F XP_005261628.1

View Full Product Details