Product Details

SNP ID
rs200503937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19720131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCGTAGAGCGCATCAGCCAGAC[C/G]GTAGAGATTCTAAAACACACGGTGG
Phenotype
MIM: 138720 MIM: 602724
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GP1BB PubMed Links

Gene Details

Gene
GP1BB
Gene Name
glycoprotein Ib platelet beta subunit
There are no transcripts associated with this gene.

Gene
SEPT5
Gene Name
septin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009939.2 585 Silent Mutation ACC,ACG T94T NP_001009939.1
NM_002688.5 585 Silent Mutation ACC,ACG T85T NP_002679.2
Gene
SEPT5-GP1BB
Gene Name
SEPT5-GP1BB readthrough
There are no transcripts associated with this gene.

View Full Product Details