Product Details

SNP ID
rs200753314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37469997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCAGCTGGGGACACCAGGGTGTA[C/T]CTGGATAGAGCAGACAATGGAGGGA
Phenotype
MIM: 602577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFNG PubMed Links

Gene Details

Gene
MFNG
Gene Name
MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166343.1 1100 Missense Mutation GAT,GGT D297G NP_001159815.1
NM_002405.3 1100 Missense Mutation GAT,GGT D311G NP_002396.2

View Full Product Details