Product Details

SNP ID
rs200118882
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36991203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAGTACTTGGAGCTCTTGGATTT[A/G]CTGTTTTCTCCATATTTCTCTTCCA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TEX33 PubMed Links

Gene Details

Gene
TEX33
Gene Name
testis expressed 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163857.1 946 Silent Mutation NP_001157329.1
NM_178552.3 946 Silent Mutation NP_848647.1
XM_011530165.2 946 Silent Mutation XP_011528467.1
XM_011530166.1 946 Silent Mutation XP_011528468.1

View Full Product Details