Product Details

SNP ID
rs201605955
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40679621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGGGTGGAGGGGAGCTCAGCTC[G/T]GTTGTGGGAGCAGGCGACCGGCACT
Phenotype
MIM: 601751
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MCHR1 PubMed Links

Gene Details

Gene
MCHR1
Gene Name
melanin concentrating hormone receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005297.3 444 Missense Mutation CGG,CTG R59L NP_005288.3

View Full Product Details