Product Details

SNP ID
rs200962268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40679648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGGAGCAGGCGACCGGCACTG[A/G]CTGGATGGACCTGGAAGCCTCGCTG
Phenotype
MIM: 601751
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCHR1 PubMed Links

Gene Details

Gene
MCHR1
Gene Name
melanin concentrating hormone receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005297.3 471 Missense Mutation GAC,GGC D68G NP_005288.3

View Full Product Details