Product Details

SNP ID
rs201027876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:16784380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCAGCGAATGGCCCCATAAGTAA[C/T]TGATAACAGGGAAAATGTCATCAGC
Phenotype
MIM: 611674
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XKR3 PubMed Links

Gene Details

Gene
XKR3
Gene Name
XK related 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318251.1 722 Missense Mutation ATT,GTT I207V NP_001305180.1
NM_175878.4 722 Missense Mutation ATT,GTT I207V NP_787074.2
XM_017028600.1 722 Missense Mutation ATT,GTT I207V XP_016884089.1

View Full Product Details