Product Details

SNP ID
rs200519385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29555961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCAGAGGCGAGATCTTCAAGG[G/T]GATCATGATCCTAGACTCCATGTGT
Phenotype
MIM: 603249 MIM: 612733
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NIPSNAP1 PubMed Links

Gene Details

Gene
NIPSNAP1
Gene Name
nipsnap homolog 1 (C. elegans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202502.1 783 Missense Mutation ACC,CCC T257P NP_001189431.1
NM_003634.3 783 Missense Mutation ACC,CCC T277P NP_003625.2
Gene
THOC5
Gene Name
THO complex 5
There are no transcripts associated with this gene.

View Full Product Details