Product Details

SNP ID
rs200345991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39350070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCGCCTTCGACCCCTACACCCT[A/G]GTCCGGCAGCCGCACACCATCCTGC
Phenotype
MIM: 603925
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYNGR1 PubMed Links

Gene Details

Gene
SYNGR1
Gene Name
synaptogyrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004711.4 122 Silent Mutation CTA,CTG L20L NP_004702.2
NM_145731.3 122 Silent Mutation CTA,CTG L20L NP_663783.1
NM_145738.2 122 Intron NP_663791.1

View Full Product Details