Product Details

SNP ID
rs201372986
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50482845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGCCTGTGGCAACTCATGGGAC[C/T]GGCCGGCCGGCAGGACTCAGCTCCT
Phenotype
MIM: 608682 MIM: 606774 MIM: 603560
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ADM2 PubMed Links

Gene Details

Gene
ADM2
Gene Name
adrenomedullin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253845.1 440 Missense Mutation CCG,CTG P130L NP_001240774.1
XM_017028940.1 440 Missense Mutation CCG,CTG P130L XP_016884429.1
Gene
MIOX
Gene Name
myo-inositol oxygenase
There are no transcripts associated with this gene.

Gene
SBF1
Gene Name
SET binding factor 1
There are no transcripts associated with this gene.

View Full Product Details