Product Details

SNP ID
rs202216099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39133930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGTCTCGGAAGAAGCCCTCAGC[C/T]GCCTGGGCCTCGCGGAAGGTGACGG
Phenotype
MIM: 608457
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CBX7 PubMed Links

Gene Details

Gene
CBX7
Gene Name
chromobox 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175709.3 750 Silent Mutation GCA,GCG A239A NP_783640.1
XM_005261413.4 750 Silent Mutation GCA,GCG A146A XP_005261470.1
XM_006724174.3 750 Silent Mutation GCA,GCG A247A XP_006724237.1
XM_006724175.3 750 Silent Mutation GCA,GCG A246A XP_006724238.1
XM_006724176.3 750 Silent Mutation GCA,GCG A188A XP_006724239.1
XM_006724177.3 750 Silent Mutation GCA,GCG A180A XP_006724240.1
XM_006724178.3 750 Silent Mutation GCA,GCG A154A XP_006724241.1
XM_011530025.2 750 Silent Mutation GCA,GCG A153A XP_011528327.1
XM_017028679.1 750 Silent Mutation GCA,GCG A238A XP_016884168.1

View Full Product Details