Product Details

SNP ID
rs202058429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:45923147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGATAGCTGCGCAGCTGTTTGAT[A/G]CGCAGGAAGGTGGGCTGCCGCAGAC
Phenotype
MIM: 601967
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WNT7B PubMed Links

Gene Details

Gene
WNT7B
Gene Name
Wnt family member 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058238.2 1133 Silent Mutation CGC,CGT R253R NP_478679.1
XM_011530366.1 1133 Silent Mutation CGC,CGT R257R XP_011528668.1

View Full Product Details