Product Details

SNP ID
rs201185401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:172102232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGTCATAGCTGCTCTTGGCGG[A/G]GTAAGGGACGTTGTTGAGGCCGCTG
Phenotype
MIM: 126255
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLX2 PubMed Links

Gene Details

Gene
DLX2
Gene Name
distal-less homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004405.3 519 Missense Mutation CCC,TCC P103S NP_004396.1
Gene
DLX2-AS1
Gene Name
DLX2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details