Product Details

SNP ID
rs200816451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:64636131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGACGTATCAATGTCAGCAAA[C/G]AGGATGTCATCCAGGGTCAAGTCTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SERTAD2 PubMed Links

Gene Details

Gene
SERTAD2
Gene Name
SERTA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014755.2 1092 Silent Mutation CTC,CTG L247L NP_055570.1
XM_005264669.2 1092 Silent Mutation CTC,CTG L247L XP_005264726.1
XM_011533204.1 1092 Silent Mutation CTC,CTG L254L XP_011531506.1
XM_011533205.2 1092 Silent Mutation CTC,CTG L247L XP_011531507.1

View Full Product Details