Product Details

SNP ID
rs200857287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99245381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGGTCCACGCAATGTCTCTGCC[C/T]GACTTCTTTGATCAGAGTCTGGTAA
Phenotype
MIM: 616547
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYG2 PubMed Links

Gene Details

Gene
LYG2
Gene Name
lysozyme g2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175735.3 375 Missense Mutation AGG,GGG R88G NP_783862.2
XM_017003751.1 375 Missense Mutation AGG,GGG R88G XP_016859240.1

View Full Product Details