Product Details

SNP ID
rs200969045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27053539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCGCACACTGCCCACCCGGCCAC[A/G]CTGGGAACGCATTCGAGACCGGCCC
Phenotype
MIM: 615900
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGBL5 PubMed Links

Gene Details

Gene
AGBL5
Gene Name
ATP/GTP binding protein like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035507.2 531 Missense Mutation CAC,CGC H118R NP_001030584.1
NM_021831.5 531 Missense Mutation CAC,CGC H118R NP_068603.4
XM_005264477.2 531 Missense Mutation CAC,CGC H118R XP_005264534.1
XM_011533011.2 531 Missense Mutation CAC,CGC H118R XP_011531313.1
XM_011533012.2 531 Missense Mutation CAC,CGC H118R XP_011531314.1
XM_011533013.2 531 Missense Mutation CAC,CGC H118R XP_011531315.1
Gene
AGBL5-AS1
Gene Name
AGBL5 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TRA-AGC8-1
Gene Name
transfer RNA-Ala (AGC) 8-1
There are no transcripts associated with this gene.

View Full Product Details