Product Details
- SNP ID
-
rs201269168
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:9406847 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATTGCAGGATTCAGGGTTTCTCA[C/G]ATGTTAATACAGAGTCAAAAGCGGT
- Phenotype
-
MIM: 603817
MIM: 607153
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ASAP2
PubMed Links
Gene Details
- Gene
- ASAP2
- Gene Name
- ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
There are no transcripts associated with this gene.
- Gene
- ITGB1BP1
- Gene Name
- integrin subunit beta 1 binding protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001319066.1 |
1323 |
Missense Mutation |
TCT,TGT |
S197C |
NP_001305995.1 |
NM_001319067.1 |
1323 |
Missense Mutation |
TCT,TGT |
S197C |
NP_001305996.1 |
NM_001319068.1 |
1323 |
Missense Mutation |
TCT,TGT |
S197C |
NP_001305997.1 |
NM_001319069.1 |
1323 |
Missense Mutation |
TCT,TGT |
S153C |
NP_001305998.1 |
NM_001319070.1 |
1323 |
Missense Mutation |
TCT,TGT |
S125C |
NP_001305999.1 |
NM_004763.4 |
1323 |
Missense Mutation |
TCT,TGT |
S197C |
NP_004754.1 |
NM_022334.4 |
1323 |
Missense Mutation |
TCT,TGT |
S147C |
NP_071729.1 |
XM_005246183.4 |
1323 |
Missense Mutation |
TCT,TGT |
S197C |
XP_005246240.1 |
XM_005246184.4 |
1323 |
Missense Mutation |
TCT,TGT |
S197C |
XP_005246241.1 |
XM_005246185.4 |
1323 |
Missense Mutation |
TCT,TGT |
S197C |
XP_005246242.1 |
XM_005246189.4 |
1323 |
Missense Mutation |
TCT,TGT |
S147C |
XP_005246246.1 |
XM_006711903.3 |
1323 |
Missense Mutation |
TCT,TGT |
S197C |
XP_006711966.1 |
XM_011510416.2 |
1323 |
Missense Mutation |
TCT,TGT |
S197C |
XP_011508718.1 |
XM_017005267.1 |
1323 |
Missense Mutation |
TCT,TGT |
S197C |
XP_016860756.1 |
XM_017005268.1 |
1323 |
Missense Mutation |
TCT,TGT |
S197C |
XP_016860757.1 |
XM_017005269.1 |
1323 |
Missense Mutation |
TCT,TGT |
S147C |
XP_016860758.1 |
XM_017005270.1 |
1323 |
Missense Mutation |
TCT,TGT |
S147C |
XP_016860759.1 |
View Full Product Details