Product Details
- SNP ID
-
rs201359857
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:207081240 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATATGTCTCTTCATGTGGAGGGC[A/G]AGATGGTCAGACCTGGAAAAACACC
- Phenotype
-
MIM: 604865
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KLF7
PubMed Links
Gene Details
- Gene
- KLF7
- Gene Name
- Kruppel like factor 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001270942.1 |
1116 |
Missense Mutation |
TCG,TTG |
S227L |
NP_001257871.1 |
NM_001270943.1 |
1116 |
Silent Mutation |
CTC,CTT |
L261L |
NP_001257872.1 |
NM_001270944.1 |
1116 |
Silent Mutation |
CTC,CTT |
L266L |
NP_001257873.1 |
NM_003709.3 |
1116 |
Silent Mutation |
CTC,CTT |
L294L |
NP_003700.1 |
XM_005246926.3 |
1116 |
Silent Mutation |
CTC,CTT |
L295L |
XP_005246983.1 |
XM_006712815.3 |
1116 |
Silent Mutation |
CTC,CTT |
L295L |
XP_006712878.1 |
XM_011512072.2 |
1116 |
Silent Mutation |
CTC,CTT |
L295L |
XP_011510374.1 |
XM_011512073.2 |
1116 |
Silent Mutation |
CTC,CTT |
L264L |
XP_011510375.1 |
XM_011512074.2 |
1116 |
Silent Mutation |
CTC,CTT |
L262L |
XP_011510376.1 |
XM_011512075.1 |
1116 |
Intron |
|
|
XP_011510377.1 |
XM_017005161.1 |
1116 |
Silent Mutation |
CTC,CTT |
L294L |
XP_016860650.1 |
XM_017005162.1 |
1116 |
Silent Mutation |
CTC,CTT |
L267L |
XP_016860651.1 |
XM_017005163.1 |
1116 |
Intron |
|
|
XP_016860652.1 |
XM_017005164.1 |
1116 |
Missense Mutation |
TCG,TTG |
S227L |
XP_016860653.1 |
View Full Product Details