Product Details

SNP ID
rs200873971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:64456309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCTTTGCAATCAGCTTGACCTG[C/T]GGGGACTCAGAAGACCCTCCTGCCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGALSL PubMed Links

Gene Details

Gene
LGALSL
Gene Name
galectin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014181.2 572 Silent Mutation TGC,TGT C73C NP_054900.2

View Full Product Details