Product Details

SNP ID
rs201014550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169701473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGTAGATAAACAGTTACAACAG[A/G]GAGTGAATTATACACAAATTGTTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC173 PubMed Links

Gene Details

Gene
CCDC173
Gene Name
coiled-coil domain containing 173
There are no transcripts associated with this gene.

Gene
PHOSPHO2
Gene Name
phosphatase, orphan 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008489.3 801 Missense Mutation AGA,GGA R168G NP_001008489.1
NM_001199285.1 801 Missense Mutation AGA,GGA R168G NP_001186214.1
NM_001199286.1 801 Missense Mutation AGA,GGA R168G NP_001186215.1
NM_001199287.1 801 Missense Mutation AGA,GGA R168G NP_001186216.1
NM_001199288.1 801 Missense Mutation AGA,GGA R168G NP_001186217.1
Gene
PHOSPHO2-KLHL23
Gene Name
PHOSPHO2-KLHL23 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199290.1 801 Intron NP_001186219.1

View Full Product Details