Product Details

SNP ID
rs201285698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:75049489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTGTCATGGTCTTGGAGTCACT[G/T]CGTGAAGAGCAGTTGGAGGTCAGGT
Phenotype
MIM: 162323
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TACR1 PubMed Links

Gene Details

Gene
TACR1
Gene Name
tachykinin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001058.3 1752 Silent Mutation CGA,CGC R389R NP_001049.1
NM_015727.2 1752 Intron NP_056542.1

View Full Product Details