Product Details

SNP ID
rs200876825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38931142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGAAGAGCACAGCTCACAGGC[C/T]GGGCCTGCCCAAGCACAAGGAAGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF33 PubMed Links

Gene Details

Gene
ARHGEF33
Gene Name
Rho guanine nucleotide exchange factor 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145451.2 481 Silent Mutation GCC,GCT A132A NP_001138923.2

View Full Product Details