Product Details
- SNP ID
-
rs200890046
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:88448444 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTGGAGGGCAGCTGGGCCCCCTG[A/G]ATCCCTAGGTGGCGGCTGCCAGGGA
- Phenotype
-
MIM: 612351
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FOXI3
PubMed Links
Gene Details
- Gene
- FOXI3
- Gene Name
- forkhead box I3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001135649.1 |
1026 |
Silent Mutation |
ATC,ATT |
I342I |
NP_001129121.1 |
View Full Product Details