Product Details

SNP ID
rs200904524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:157258109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACAGGATCCGAAAGTTTTTCCG[A/C]GGAAGTGGGCGAGTCTTGGCATTTA
Phenotype
MIM: 615129
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GALNT5 PubMed Links

Gene Details

Gene
GALNT5
Gene Name
polypeptide N-acetylgalactosaminyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017003237.1 562 Silent Mutation CGA,CGC R9R XP_016858726.1
XM_017003238.1 562 Silent Mutation CGA,CGC R9R XP_016858727.1

View Full Product Details