Product Details

SNP ID
rs201016476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202884512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTGAGGAAACTGCCTCCATGTG[G/T]CCAGAGAGGGTCACTATGGGAGTCT
Phenotype
MIM: 616620
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WDR12 PubMed Links

Gene Details

Gene
WDR12
Gene Name
WD repeat domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018256.3 1392 Silent Mutation GGA,GGC G255G NP_060726.3
XM_011511469.2 1392 Silent Mutation GGA,GGC G117G XP_011509771.1

View Full Product Details