Product Details

SNP ID
rs201273235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88698539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACAATTGTCCATGCTGTGCAGC[A/G]AATAGGTATGCTCTCTCACTGTCTA
Phenotype
MIM: 180430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPIA PubMed Links

Gene Details

Gene
RPIA
Gene Name
ribose 5-phosphate isomerase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144563.2 382 Missense Mutation CAA,CGA Q114R NP_653164.2

View Full Product Details