Product Details

SNP ID
rs200657890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27137376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCCCTGAGGCCCTGCTTCCTT[C/T]GGCCTTTTCTCGGGCAGCAGATGCA
Phenotype
MIM: 612195 MIM: 606395
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD1 PubMed Links

Gene Details

Gene
ABHD1
Gene Name
abhydrolase domain containing 1
There are no transcripts associated with this gene.

Gene
PREB
Gene Name
prolactin regulatory element binding
There are no transcripts associated with this gene.

Gene
PRR30
Gene Name
proline rich 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178553.3 1534 Silent Mutation CCA,CCG P318P NP_848648.2

View Full Product Details