Product Details

SNP ID
rs200726804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178490673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCATCATAGTTCATTCAGCAGAC[A/C]ACACAAGAATGGAATTAATCATTCC
Phenotype
MIM: 607774
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLEKHA3 PubMed Links

Gene Details

Gene
PLEKHA3
Gene Name
pleckstrin homology domain containing A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019091.3 679 Missense Mutation AAC,CAC N58H NP_061964.3
XM_011511689.2 679 UTR 5 XP_011509991.1

View Full Product Details