Product Details

SNP ID
rs200947523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:195738087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAATGGAGCAACATAACTTGGC[C/T]GTTTTGGTATATCTGCCCTCTTACA
Phenotype
MIM: 610061 MIM: 608733
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAH7 PubMed Links

Gene Details

Gene
DNAH7
Gene Name
dynein axonemal heavy chain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018897.2 6406 Missense Mutation CAG,CGG Q3970R NP_061720.2
XM_011511487.2 6406 Missense Mutation CAG,CGG Q3970R XP_011509789.1
XM_011511488.2 6406 Missense Mutation CAG,CGG Q3930R XP_011509790.1
XM_011511489.2 6406 Missense Mutation CAG,CGG Q3924R XP_011509791.1
XM_011511490.2 6406 Missense Mutation CAG,CGG Q3895R XP_011509792.1
XM_011511491.2 6406 Intron XP_011509793.1
XM_011511492.2 6406 Intron XP_011509794.1
XM_011511493.2 6406 Intron XP_011509795.1
XM_011511494.2 6406 Intron XP_011509796.1
XM_011511495.2 6406 Intron XP_011509797.1
XM_011511497.2 6406 Missense Mutation CAG,CGG Q2094R XP_011509799.1
XM_017004504.1 6406 Missense Mutation CAG,CGG Q3879R XP_016859993.1
Gene
SLC39A10
Gene Name
solute carrier family 39 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127257.1 6406 Intron NP_001120729.1
NM_020342.2 6406 Intron NP_065075.1
XM_005246689.4 6406 Intron XP_005246746.2
XM_011511504.2 6406 Intron XP_011509806.1
XM_011511505.2 6406 Intron XP_011509807.1
XM_011511506.2 6406 Intron XP_011509808.1
XM_011511507.2 6406 Intron XP_011509809.1
XM_017004522.1 6406 Intron XP_016860011.1
XM_017004523.1 6406 Intron XP_016860012.1

View Full Product Details