Product Details

SNP ID
rs201058038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:98796202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCCCCTGCTTCCGGTCCACAGC[G/T]GGCTTCACAGGGGTTATCAGGAAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1211L PubMed Links

Gene Details

Gene
KIAA1211L
Gene Name
KIAA1211 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207362.2 2961 Silent Mutation NP_997245.2
XM_005263923.3 2961 Silent Mutation XP_005263980.1
XM_005263924.3 2961 Silent Mutation XP_005263981.1
XM_005263926.3 2961 Silent Mutation XP_005263983.1
XM_011511093.1 2961 Silent Mutation XP_011509395.1
XM_011511094.1 2961 Silent Mutation XP_011509396.1
XM_011511095.2 2961 Silent Mutation XP_011509397.1
XM_011511096.1 2961 Silent Mutation XP_011509398.1
XM_017003971.1 2961 Silent Mutation XP_016859460.1
XM_017003972.1 2961 Silent Mutation XP_016859461.1
XM_017003973.1 2961 Intron XP_016859462.1

View Full Product Details