Product Details

SNP ID
rs200672180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:97657953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAGCCACAGTACCTTTGAAAAG[C/T]GTTGAGCCACCTGAGAGCACGATGT
Phenotype
MIM: 605144 MIM: 123866
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTR1B PubMed Links

Gene Details

Gene
ACTR1B
Gene Name
ARP1 actin-related protein 1 homolog B, centractin beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005735.3 1177 Silent Mutation ACA,ACG T305T NP_005726.1
XM_005263854.4 1177 Silent Mutation ACA,ACG T231T XP_005263911.1
XM_017003116.1 1177 Silent Mutation ACA,ACG T261T XP_016858605.1
Gene
COX5B
Gene Name
cytochrome c oxidase subunit 5B
There are no transcripts associated with this gene.

View Full Product Details