Product Details

SNP ID
rs201087297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219374924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCGGATAGAGTGCATGGCCAG[C/T]TGGGGGCTGCCTAAATCCAGCACCC
Phenotype
MIM: 611367
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNPEP PubMed Links

Gene Details

Gene
DNPEP
Gene Name
aspartyl aminopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319116.1 1376 Silent Mutation CAA,CAG Q454Q NP_001306045.1
NM_001319117.1 1376 Silent Mutation CAA,CAG Q418Q NP_001306046.1
NM_001319118.1 1376 Silent Mutation CAA,CAG Q432Q NP_001306047.1
NM_001319119.1 1376 Silent Mutation CAA,CAG Q432Q NP_001306048.1
NM_001319120.1 1376 Silent Mutation CAA,CAG Q410Q NP_001306049.1
NM_001319121.1 1376 Silent Mutation CAA,CAG Q393Q NP_001306050.1
NM_001319122.1 1376 Silent Mutation CAA,CAG Q393Q NP_001306051.1
NM_012100.3 1376 Silent Mutation CAA,CAG Q446Q NP_036232.2

View Full Product Details