Product Details

SNP ID
rs201248838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73239684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGAAAGAGGGGACTGATAGCTC[C/G]CAAGGCATCCCCCAGCTTGTGAGTA
Phenotype
MIM: 605140
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCT7 PubMed Links

Gene Details

Gene
CCT7
Gene Name
chaperonin containing TCP1 subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009570.2 191 Intron NP_001009570.1
NM_001166284.1 191 Intron NP_001159756.1
NM_001166285.1 191 UTR 5 NP_001159757.1
NM_006429.3 191 Silent Mutation TCC,TCG S16S NP_006420.1
XM_011532478.2 191 UTR 5 XP_011530780.1
XM_011532479.1 191 UTR 5 XP_011530781.1
Gene
PRADC1
Gene Name
protease associated domain containing 1
There are no transcripts associated with this gene.

View Full Product Details