Product Details

SNP ID
rs200949260
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27577626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGATTCTGCTTCAGGGATGACA[C/T]CTGGGTTAGGACATCGAGTTCCTGA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C2orf16 PubMed Links

Gene Details

Gene
C2orf16
Gene Name
chromosome 2 open reading frame 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032266.3 1105 Missense Mutation CCT,TCT P352S NP_115642.3
Gene
ZNF512
Gene Name
zinc finger protein 512
There are no transcripts associated with this gene.

View Full Product Details