Product Details

SNP ID
rs201297775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96326859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGCAGAAGGCAGAGAACTTCTG[A/G]ACAGGAGACACATCCAGTGACCAGT
Phenotype
MIM: 602332 MIM: 601664
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITPRIPL1 PubMed Links

Gene Details

Gene
ITPRIPL1
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008949.2 563 Missense Mutation TGA,TGG *76W NP_001008949.1
NM_001163523.1 563 Missense Mutation TGA,TGG *68W NP_001156995.1
NM_001163524.1 563 Missense Mutation TGA,TGG *68W NP_001156996.1
NM_001324490.1 563 Missense Mutation TGA,TGG *68W NP_001311419.1
NM_178495.5 563 Missense Mutation TGA,TGG *84W NP_848590.3
XM_017003427.1 563 Nonsense Mutation TGA,TGG *76W XP_016858916.1
Gene
LOC105373495
Gene Name
uncharacterized LOC105373495
There are no transcripts associated with this gene.

Gene
NCAPH
Gene Name
non-SMC condensin I complex subunit H
There are no transcripts associated with this gene.

Gene
SNRNP200
Gene Name
small nuclear ribonucleoprotein U5 subunit 200
There are no transcripts associated with this gene.

View Full Product Details