Product Details

SNP ID
rs200605481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26310814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTAAGAATGAGCAGGGCTTTGA[C/T]CACCCCCAGCAGAGCTTGGCGCTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRF3 PubMed Links

Gene Details

Gene
ADGRF3
Gene Name
adhesion G protein-coupled receptor F3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145168.1 2760 Missense Mutation NP_001138640.1
NM_001145169.1 2760 Missense Mutation NP_001138641.1
NM_001321971.1 2760 Missense Mutation NP_001308900.1
NM_001321975.1 2760 Missense Mutation NP_001308904.1
NM_153835.3 2760 Missense Mutation NP_722577.2
XM_011532621.2 2760 Missense Mutation XP_011530923.1
XM_011532622.2 2760 Missense Mutation XP_011530924.1
XM_011532623.2 2760 Missense Mutation XP_011530925.1
XM_017003505.1 2760 Missense Mutation XP_016858994.1
XM_017003506.1 2760 Missense Mutation XP_016858995.1

View Full Product Details