Product Details

SNP ID
rs200756266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:75049497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTCTTGGAGTCACTTCGTGAAG[A/T]GCAGTTGGAGGTCAGGTCCAGGGAC
Phenotype
MIM: 162323
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TACR1 PubMed Links

Gene Details

Gene
TACR1
Gene Name
tachykinin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001058.3 1744 Missense Mutation ACT,TCT T387S NP_001049.1
NM_015727.2 1744 Intron NP_056542.1

View Full Product Details